Merck
  • Cell-Mediated Immunity Generated in Response to a Purified Inactivated Vaccine for Dengue Virus Type 1.
Cell-Mediated Immunity Generated in Response to a Purified Inactivated Vaccine for Dengue Virus Type 1.
mSphere (2020-01-24)
Heather Friberg, Luis J Martinez, Leyi Lin, Jason M Blaylock, Rafael A De La Barrera, Alan L Rothman, J Robert Putnak, Kenneth H Eckels, Stephen J Thomas, Richard G Jarman, Jeffrey R Currier
ABSTRACT
Dengue is the most prevalent arboviral disease afflicting humans, and a vaccine appears to be the most rational means of control. Dengue vaccine development is in a critical phase, with the first vaccine licensed in some countries where dengue is endemic but demonstrating insufficient efficacy in immunologically naive populations. Since virus-neutralizing antibodies do not invariably correlate with vaccine efficacy, other markers that may predict protection, including cell-mediated immunity, are urgently needed. Previously, the Walter Reed Army Institute of Research developed a monovalent purified inactivated virus (PIV) vaccine candidate against dengue virus serotype 1 (DENV-1) adjuvanted with alum. The PIV vaccine was safe and immunogenic in a phase I dose escalation trial in healthy, flavivirus-naive adults in the United States. From that trial, peripheral blood mononuclear cells obtained at various time points pre- and postvaccination were used to measure DENV-1-specific T cell responses. After vaccination, a predominant CD4+ T cell-mediated response to peptide pools covering the DENV-1 structural proteins was observed. Over half (13/20) of the subjects produced interleukin-2 (IL-2) in response to DENV peptides, and the majority (17/20) demonstrated peptide-specific CD4+ T cell proliferation. In addition, analysis of postvaccination cell culture supernatants demonstrated an increased rate of production of cytokines, including gamma interferon (IFN-γ), IL-5, and granulocyte-macrophage colony-stimulating factor (GM-CSF). Overall, the vaccine was found to have elicited DENV-specific CD4+ T cell responses as measured by enzyme-linked immunosorbent spot (ELISpot), intracellular cytokine staining (ICS), lymphocyte proliferation, and cytokine production assays. Thus, together with antibody readouts, the use of a multifaceted measurement of cell-mediated immune responses after vaccination is a useful strategy for more comprehensively characterizing immunity generated by dengue vaccines.IMPORTANCE Dengue is a tropical disease transmitted by mosquitoes, and nearly half of the world's population lives in areas where individuals are at risk of infection. Several vaccines for dengue are in development, including one which was recently licensed in several countries, although its utility is limited to people who have already been infected with one of the four dengue viruses. One major hurdle to understanding whether a dengue vaccine will work for everyone-before exposure-is the necessity of knowing which marker can be measured in the blood to signal that the individual has protective immunity. This report describes an approach measuring multiple different parts of immunity in order to characterize which signals one candidate vaccine imparted to a small number of human volunteers. This approach was designed to be able to be applied to any dengue vaccine study so that the data can be compared and used to inform future vaccine design and/or optimization strategies.
Product Description
Millipore
Multiscreen® 96 well Plate, hydrophilic PVDF membrane, pore size 1.2 μm, sterile
Pricing and availability is not currently available.