Merck
  • Use of Transabdominal Ultrasound for the detection of intra-peritoneal tumor engraftment and growth in mouse xenografts of epithelial ovarian cancer.
Use of Transabdominal Ultrasound for the detection of intra-peritoneal tumor engraftment and growth in mouse xenografts of epithelial ovarian cancer.
PloS one (2020-04-30)
Laura M Chambers, Emily Esakov, Chad Braley, Mariam AlHilli, Chad Michener, Ofer Reizes
ABSTRACT
To evaluate intraperitoneal (IP) tumor engraftment, metastasis and growth in a pre-clinical murine epithelial ovarian cancer (EOC) model using both transabdominal ultrasound (TAUS) and bioluminescence in vivo imaging system (IVIS). Ten female C57Bl/6J mice at six weeks of age were included in this study. Five mice underwent IP injection of 5x106 ID8-luc cells (+ D- luciferin) and the remaining five mice underwent IP injection of ID8-VEGF cells. Monitoring of tumor growth and ascites was performed weekly starting at seven days post-injection until study endpoint. ID8-luc mice were monitored using both TAUS and IVIS, and ID8-VEGF mice underwent TAUS monitoring only. Individual tumor implant dimension and total tumor volume were calculated. Average luminescent intensity was calculated and reported per mouse abdomen. Tumor detection was confirmed by gross evaluation and histopathology. All data are presented as mean +/- standard deviation. Overall, tumors were successfully detected in all ten mice using TAUS and IVIS, and tumor detection correlated with terminal endpoint histology/ H&E staining. For TAUS, the smallest confirmed tumor measurements were at seven days post-injection with mean long axis of 2.23mm and mean tumor volume of 4.17mm3. However, IVIS imaging was able to detect tumor growth at 14 days post-injection. Ascites formation was detected in mice at 21 days post-injection. TAUS is highly discriminatory for monitoring EOC in pre-clinical murine model, allowing for detection of tumor dimension as small as 2 mm and as early as seven days post-injection compared to IVIS. In addition, TAUS provides relevant information for ascites development and detection of multiple small metastatic tumor implants. TAUS provides an accurate and reliable method to detect and monitor IP EOC growth in mouse xenografts.
Product Description
Millipore
Millipore® Steriflip® Vacuum Tube Top Filter, pore size 0.45 μm, PVDF membrane, funnel capacity 50 mL, pack of 25 ea
Pricing and availability is not currently available.