Merck
  • Catalyst control in sequential asymmetric allylic substitution: stereodivergent access to N,N-diprotected unnatural amino acids.
Catalyst control in sequential asymmetric allylic substitution: stereodivergent access to N,N-diprotected unnatural amino acids.
The Journal of organic chemistry (2011-05-14)
Paolo Tosatti, Amanda J Campbell, David House, Adam Nelson, Stephen P Marsden
ABSTRACT
The sequential use of Cu-catalyzed asymmetric allylic alkylation, olefin cross-metathesis, and Ir-catalyzed asymmetric allylic amination allows the concise, stereodivergent synthesis of complex chiral amines with complete regiocontrol and good diastereoselectivity, exemplified by the synthesis of a pair of diastereoisomeric unnatural branched amino acid derivatives.
Sigma-Aldrich
QuadraPure® TU, macroporous, 400-600 μm particle size
Pricing and availability is not currently available.