Merck
Role of C-mannosylation in the secretion of mindin.
Biochimica et biophysica acta. General subjects (2020-05-18)
Yoko Inai, Kana Ueda, In-Sook Lee Matsui, Michiko Tajiri, Shiho Minakata, Yoshinao Wada, Yoshito Ihara
ABSTRACT
Mindin (spondin2), a secretory protein related to neural development and immunity, is a member of thrombospondin type I repeat (TSR) superfamily proteins, and has a unique glycosylation of C-mannosylation in its structure. However, it remains unclear whether C-mannosylation plays a functional role in the biosynthesis of mindin in cells. Protein C-mannosylation was analyzed by mass spectrometry. Mindin expression was examined by immunoblot and immunofluorescence analyses in COS-7 cells transfected with the expression vectors for wild type (mindin-WT) or C-mannosylation-defective mutant of mindin (mindin-mutF). The redox status was examined in mindin by using 4-acetoamide-4'-maleimidylstilbene-2,2'-disulfonate. When mindin cDNA was expressed in COS-7 cells, C-mannosylation of mindin was confirmed at Trp257 by mass spectrometry. In cells expressing a mindin-mutF, secretion of the mutant was significantly inhibited compared with mindin-WT. In immunofluorescence analysis, mindin-mutF was accumulated in the endoplasmic reticulum (ER), whereas mindin-WT was detected in the Golgi. In addition, mindin-mutF showed an enhanced interaction with calreticulin, an ER-resident chaperone, in cells. In cells, reduced forms were increased in mindin-mutF, compared with a mostly oxidized form of mindin-WT. In the presence of chemical chaperones such as dimethylsulfoxide or 4-phenylbutyrate, inhibited secretion of mindin-mutF was ameliorated in cells, although redox-dependent folding was not affected. C-Mannosylation of mindin facilitates its secretion especially through modulating disulfide bond formation in mindin in cells. These results suggest that C-mannosylation plays a functional role in the redox-dependent folding and transport of TSR superfamily proteins in cells.
Product Description
Supelco
N-(Propionyloxy)succinimide, for LC-MS derivatization, LiChropur, ≥95% (HPLC)
Pricing and availability is not currently available.