Merck
  • Phytochemical Screening and Acanthamoebic Activity of Shoots from in Vitro Cultures and in Vivo Plants of Eryngium alpinum L.-The Endangered and Protected Species.
Phytochemical Screening and Acanthamoebic Activity of Shoots from in Vitro Cultures and in Vivo Plants of Eryngium alpinum L.-The Endangered and Protected Species.
Molecules (Basel, Switzerland) (2020-04-05)
Małgorzata Kikowska, Dariusz Kruszka, Monika Derda, Edward Hadaś, Barbara Thiem
ABSTRACT
Genetically uniform shoots of Eryngium alpinum L. cultured in vitro were subjected to the qualitative analysis applying the UPLC-HESI-HRMS technique. In vitro cultures give the opportunity to perform the phytochemical studies on the protected species without harvesting the plant material from the natural environment. The phytochemical screening of the crude methanolic extracts of shoots, both from in vitro cultures and in vivo plants, revealed the presence of phenolic acids, coumarins, flavonoids, triterpenoid saponins, amino acids, or dipeptides. Active compounds detected are known to have medicinal importance, and for this reason, the present study represents a preliminary investigation of the extracts against pathogenic and opportunistic amoeba. Among the extracts tested, the extract of shoots from in vitro cultures exhibited remarkable amoebicidal action against trophozoites. On the second day of treatment, the extract at the concentrations of 5 mg/mL, 2.5 mg/mL, and 0.5 mg/mL showed the highest antiamoebicidal effect: the inhibition of trophozoites reached 81.14%, 66.38%, and 54.99%, respectively. To our best knowledge, the present report is the first to show the phytochemical screening and to discuss the antiamoebic activity of Eryngium alpinum L. shoots, both from in vitro cultures and in vivo plants.
Product Description
Ethanol solution, NIST® SRM® 2899a, nominal 25% by mass