Merck
  • Solid-phase micro extraction of food irradiation marker 2-dodecylcyclobutanone (2-DCB) from chicken jerky treated with glycerol.
Solid-phase micro extraction of food irradiation marker 2-dodecylcyclobutanone (2-DCB) from chicken jerky treated with glycerol.
Journal of food science (2020-07-08)
Mostafa Taghvaei, Christopher Sommers, Olgica Ceric, Faris Hussain, Umut Yucel, J Scott Smith
ABSTRACT
The food irradiation marker, 2-dodecylcyclobutanone (2-DCB), assayed by SPME provides a fast and simple method to estimate the irradiation history of fat-containing food products. The SPME conditions were optimized to maximize the extraction of 2-DCB from chicken jerky treats (CJT) irradiated at low (5 kGy) and high (50 kGy) doses. The extracted 2-DCB was measured using GC-MS in selected ion mode (m/z 98, and 112). Water dilution (1:5) was needed to mobilize 2-DCB and allow partition to the headspace form the CJT matrix. Increasing the incubation temperature to 80 °C resulted in higher response. Spiking control jerky samples with 2-DCB from 10 to 150 ng/g CJT compared with spiking water revealed a significant food matrix effect. This method provides a fast, simple, and environmental friendly alternative for the existing solvent extraction methods.
Product Description
Supelco
2-Dodecylcyclobutanone, analytical standard
Pricing and availability is not currently available.
Sigma-Aldrich
Poly(dimethylsiloxane), viscosity 1.0 cSt (25 °C)
Pricing and availability is not currently available.