Merck
  • The effect of baicalein-loaded Y-shaped miktoarm copolymer on spatial memory and hippocampal expression of DHCR24, SELADIN and SIRT6 genes in rat model of Alzheimer.
The effect of baicalein-loaded Y-shaped miktoarm copolymer on spatial memory and hippocampal expression of DHCR24, SELADIN and SIRT6 genes in rat model of Alzheimer.
International journal of pharmaceutics (2020-06-17)
Mozhgan Aghajanzadeh, Sina Andalib, Hossein Danafar, Kobra Rostamizadeh, Ali Sharafi
ABSTRACT
In the present study, we successfully synthesized nanocarriers (NCs) based on Y-shaped miktoarm copolymers, Poly Ethylene Glycol-Lysine-(Poly Caprolactone)2 (PEG-Lys-PCL2), which were loaded by baicalein (B) through the nanoprecipitation process to assess their in-vitro and in-vivo properties. We applied various methods and measurements including proton nuclear magnetic resonance (HNMR), dynamic light scattering (DLS), differential scanning calorimetry (DSC), Fourier-transform infrared spectroscopy (FTIR), MTT assay, hemolysis test, lethal dose, real-time PCR, and Morris water maze. The results of DLS indicated that the size and zeta potential of the obtained NCs and B-loaded NCs were acceptable. Also, in-vivo and in-vitro biocompatibility examinations proved that miktoarm-based NCs were safe, and all rats treated with miktoarm-based NCs did not exhibit any remarkable weight loss during the experiment. The results of the Morris water maze (in-vivo test) revealed that the normal saline-treated group, as well as B-miktoarm + Scopolamine (M + B + S) and B-miktoarm-Tween80 + Scopolamine (M + B + T + S) pretreatment groups, spent more time in the target quadrant. Thus, this experiment showed that pretreatment of rats with M + B + S and M + B + T + S had the most effects on spatial memory. According to quantitative PCR analysis, we hypothesized that, in comparison with other experimental groups, pretreatment of rats with M + B + T + S could be more effective in preventing cholinergic dysfunction, brain oxidative stress and cognitive deficits which cause by Scopolamine HBr. This outcome may be partially due to the upregulation of DHCR24, SELADIN, and SIRT6 in entire of the hippocampal region of normal saline-treated and M + B + T + S pretreatment groups. These results may be because mimicking the cell membrane structure would be an excellent feature for miktoarm, and partial coating of Tween-80 can play a critical role for PEG-Lys-PCL2-based NCs in crossing the brain cell membrane, and they can easily be uptaken by the cells. Eventually, all of the obtained data confirmed that PEG-Lys-PCL2 miktoarm star copolymers are suitable for delivering therapeutic agents to the brain for the treatment of Alzheimer's disease (AD). Also, it seems that baicalein should be taken into account as a potent compound for the treatment of AD.
Sigma-Aldrich
N,N′-Dicyclohexylurea, 98%
Pricing and availability is not currently available.