Merck
  • Dual stimuli-responsive release of aptamer AS1411 decorated erlotinib loaded chitosan nanoparticles for non-small-cell lung carcinoma therapy.
Dual stimuli-responsive release of aptamer AS1411 decorated erlotinib loaded chitosan nanoparticles for non-small-cell lung carcinoma therapy.
Carbohydrate polymers (2020-07-29)
Kandasamy Saravanakumar, Anbazhagan Sathiyaseelan, Arokia Vijaya Anand Mariadoss, Elango Jeevithan, Xiaowen Hu, Sukjin Shin, Myeong-Hyeon Wang
ABSTRACT
The present work was developed the pH dependent-aptamer AS1411 (APT) decorated and erlotinib (En) loaded chitosan nanoparticles (CSNPs) for promising non-small-cell lung carcinoma (NSCLC) treatment. The characterization studies revealed that formulated APT-En-CSNPs were spherical in shape with size of 165.95 d. nm and PDI of 0.212. FTIR spectrum recorded molecular chemical interactions with composition of En or En-CSNPs. Cell viability assay, flow cytometry and fluorescent microscopy results revealed that APT-En-CSNPs triggered cancer cell death through pH-sensitive and nucleolin receptor-targeted release of En. The decoration of the APT improved the cellular uptake of En as evidenced by cellular sensing fluorescence and BioTEM assay. The APT-En-CSNPs induced the apoptosis through excessive ROS generation, nucleus damage and Δψm loss in the A549 cells. Hence, the present study revealed that the APT-En-CSNPs improved the therapeutic efficiency of En in NSCLC through the nucleolin targeted drug release.
Sigma-Aldrich
Erlotinib hydrochloride, ≥98% (HPLC)