Merck
  • MiR-92a regulates brown adipocytes differentiation, mitochondrial oxidative respiration, and heat generation by targeting SMAD7.
MiR-92a regulates brown adipocytes differentiation, mitochondrial oxidative respiration, and heat generation by targeting SMAD7.
Journal of cellular biochemistry (2019-11-07)
Zhipin Zhang, Huixin Jiang, Xiang Li, Xiaomin Chen, Yihua Huang
ABSTRACT
Brown adipocytes are rich in mitochondria and linked to the body's blood fat levels and obesity. MiR-92a is negatively correlated with the activity of brown adipocytes. This study aimed to explore the mechanism of miR-92a on brown adipocytes. The expression of miR-92a in C2C12 cell was detected by a quantitative real-time-polymerase chain reaction (qRT-PCR). C2C12 cells were induced to brown adipocytes. The direct target gene of miR-92a was determined using the dual-luciferase reporter assay. Brown adipocytes were treated with isoprenaline (Iso) and transfected by miR-92a inhibitor and siSMAD7. The expression of heat-producing genes and adipose differentiation genes related to brown adipocytes were detected by qRT-PCR and Western blot analysis. The expression of SMAD7, p-SMAD2, and p-SMAD3 were detected using Western blot analysis. The mitochondrial content was measured by mitotracker fluorescent staining. MiR-92a inhibitor significantly decreased the expression of miR-92a in C2C12 cells. MiR-92a inhibitor could upregulate the expression of Ucp1, Cox7a1, Elovl3, Ppargc1α, PPARγ, and FABP4, and its effect on Ucp1 was increased after the treatment of isoprenaline. Moreover, miR-92a inhibitor increased mitochondrial content, oxygen consumption rate (OCR) and the expression of SMAD7 and suppressed the expressions of p-SMAD2 and p-SMAD3, whereas miR-92a directly targeted SMAD7 to exert its inhibitory effects. SiSMAD7 reversed the effects of the inhibitor on heat-producing genes, mitochondrial content, OCR and the expressions of SMAD7, p-SMAD2, and p-SMAD3 in brown adipocytes. Blocking miR-92a might promote brown adipocytes differentiation, mitochondrial oxidative respiration, and thermogenesis by targeting SMAD7 to inhibit the expressions of p-SMAD2 and p-SMAD3.
Sigma-Aldrich
Dimethyl sulfoxide-d6, "100%", 99.96 atom % D
Sigma-Aldrich
3-Isobutyl-1-methylxanthine, ≥99%, BioUltra
Sigma-Aldrich
Carbonyl cyanide 4-(trifluoromethoxy)phenylhydrazone, ≥98% (TLC), powder
Sigma-Aldrich
Indomethacin, 98.5-100.5% (in accordance with EP)
Sigma-Aldrich
Rosiglitazone, ≥98% (HPLC)
Sigma-Aldrich
Dexamethasone, ≥98% (HPLC), powder