Merck
Genetic manipulation of LKB1 elicits lethal metastatic prostate cancer.
The Journal of experimental medicine (2020-03-29)
Ivana Hermanova, Patricia Zúñiga-García, Alfredo Caro-Maldonado, Sonia Fernandez-Ruiz, Fernando Salvador, Natalia Martín-Martín, Amaia Zabala-Letona, Marc Nuñez-Olle, Verónica Torrano, Laura Camacho, Jose M Lizcano, Ana Talamillo, Suzanne Carreira, Bora Gurel, Ana R Cortazar, Marc Guiu, Jose I López, Anabel Martinez-Romero, Ianire Astobiza, Lorea Valcarcel-Jimenez, Mar Lorente, Amaia Arruabarrena-Aristorena, Guillermo Velasco, Antonio Gomez-Muñoz, Cristian Suárez-Cabrera, Iris Lodewijk, Juana M Flores, James D Sutherland, Rosa Barrio, Johann S de Bono, Jesús M Paramio, Jan Trka, Mariona Graupera, Roger R Gomis, Arkaitz Carracedo
ABSTRACT
Gene dosage is a key defining factor to understand cancer pathogenesis and progression, which requires the development of experimental models that aid better deconstruction of the disease. Here, we model an aggressive form of prostate cancer and show the unconventional association of LKB1 dosage to prostate tumorigenesis. Whereas loss of Lkb1 alone in the murine prostate epithelium was inconsequential for tumorigenesis, its combination with an oncogenic insult, illustrated by Pten heterozygosity, elicited lethal metastatic prostate cancer. Despite the low frequency of LKB1 deletion in patients, this event was significantly enriched in lung metastasis. Modeling the role of LKB1 in cellular systems revealed that the residual activity retained in a reported kinase-dead form, LKB1K78I, was sufficient to hamper tumor aggressiveness and metastatic dissemination. Our data suggest that prostate cells can function normally with low activity of LKB1, whereas its complete absence influences prostate cancer pathogenesis and dissemination.
Sigma-Aldrich
Methyl cellulose, viscosity: 1,500 cP
Pricing and availability is not currently available.