Merck
  • Doublecortin-like kinase 1 promotes hepatocyte clonogenicity and oncogenic programming via non-canonical β-catenin-dependent mechanism.
Doublecortin-like kinase 1 promotes hepatocyte clonogenicity and oncogenic programming via non-canonical β-catenin-dependent mechanism.
Scientific reports (2020-07-01)
Naushad Ali, Charles B Nguyen, Parthasarathy Chandrakesan, Roman F Wolf, Dongfeng Qu, Randal May, Tatiana Goretsky, Javid Fazili, Terrence A Barrett, Min Li, Mark M Huycke, Michael S Bronze, Courtney W Houchen
ABSTRACT
Chronic liver injury is a risk factor for cirrhosis and hepatocellular carcinoma (HCC). The molecular mechanisms that regulate the decision between normal injury repair and neoplastic initiation are unclear. Doublecortin-like kinase 1 (DCLK1), a tumor stem cell marker, is induced during cirrhosis and HCC. Here, we demonstrate that DCLK1-overexpressing primary human hepatocytes formed spheroids in suspension cultures. Spheroids derived from DCLK1-overexpressing hepatoma cells showed high level expression of active β-catenin, α-fetoprotein, and SOX9, suggesting that DCLK1 overexpression induces clonogenicity and dedifferentiated phenotypes in hepatoma cells. DCLK1 overexpression in hepatoma cells also increased phosphorylation of GSK-3β at Ser9. This was associated with an induction of a 48-kDa active β-catenin with a preserved hypophosphorylated N-terminus that interacted with nuclear TCF-4 resulting in luciferase reporter activity and cyclin D1 expression. DCLK1 downregulation inhibited 48-kDa β-catenin expression. The proteasome inhibitor bortezomib did not block the 48-kDa β-catenin, instead, caused a threefold accumulation, suggesting a proteasome-independent mechanism. Liver tissues from patients with cirrhosis and HCC revealed epithelial co-staining of DCLK1 and active β-catenin, and cleaved E-cadherin. Repopulated DCLK1-overexpressing primary human hepatocytes in humanized FRG mouse livers demonstrated active β-catenin. In conclusion, DCLK1 regulates oncogenic signaling and clonogenicity of hepatocytes by a novel non-canonical/atypical β-catenin-dependent mechanism.
Sigma-Aldrich
Anti-Active-β-Catenin (Anti-ABC) Antibody, clone 8E7, clone 8E7, Upstate®, from mouse
Pricing and availability is not currently available.
Sigma-Aldrich
Rabbit anti-Beta-catenin Antibody, Affinity Purified, Powered by Bethyl Laboratories, Inc.
Pricing and availability is not currently available.