Merck
  • Melatonin Suppresses the Expression of 45S Preribosomal RNA and Upstream Binding Factor and Enhances the Antitumor Activity of Puromycin in MDA-MB-231 Breast Cancer Cells.
Melatonin Suppresses the Expression of 45S Preribosomal RNA and Upstream Binding Factor and Enhances the Antitumor Activity of Puromycin in MDA-MB-231 Breast Cancer Cells.
Evidence-based complementary and alternative medicine : eCAM (2013-05-22)
Ji Hoon Jung, Eun Jung Sohn, Eun Ah Shin, Duckgue Lee, Bonglee Kim, Deok-Beom Jung, Ji-Hyun Kim, Miyong Yun, Hyo-Jeong Lee, Yong Koo Park, Sung-Hoon Kim
ABSTRACT
Since the dysregulation of ribosome biogenesis is closely associated with tumor progression, in the current study, the critical role of ribosome biogenesis related signaling was investigated in melatonin and/or puromycin induced apoptosis in MDA-MB-231 breast cancer cells. Despite its weak cytotoxicity, melatonin from 3 mM attenuated the expression of 45S pre-ribosomal RNA (pre-rRNA), UBF as a nucleolar transcription factor, and fibrillarin at mRNA level and consistently downregulated nucleolar proteins such as UBF and fibrillarin at protein level in MDA-MB-231 cells. Furthermore, immunofluorescence assay revealed that UBF was also degraded by melatonin in MDA-MB-231 cells. In contrast, melatonin attenuated the expression of survival genes such as Bcl-xL, Mcl-1, cyclinD1, and cyclin E, suppressed the phosphorylation of AKT, mTOR, and STAT3, and cleaved PARP and activated caspase 3 only at a high concentration of 12 mM. However, combined treatment of melatonin (3 mM) and puromycin (1 μM) synergistically inhibited viability, attenuated the expression of 45S pre-rRNA and UBF, and consistently downregulated UBF, XPO1 and IPO7, procaspase 3, and Bcl-xL in MDA-MB 231 cells. Overall, these findings suggest that melatonin can be a cancer preventive agent by combination with puromycin via the inhibition of 45S pre-rRNA and UBF in MDA-MB 231 breast cancer cells.
Sigma-Aldrich
Puromycin dihydrochloride from Streptomyces alboniger, ≥98% (HPLC), powder
Pricing and availability is not currently available.
Sigma-Aldrich
Puromycin ready made solution, 10 mg/mL in H2O, ≥98% (HPLC), Cell culture tested
Pricing and availability is not currently available.