Merck
Phosphatidic Acid and Cardiolipin Coordinate Mitochondrial Dynamics.
Trends in cell biology (2017-09-16)
Shoichiro Kameoka, Yoshihiro Adachi, Koji Okamoto, Miho Iijima, Hiromi Sesaki
ABSTRACT
Membrane organelles comprise both proteins and lipids. Remodeling of these membrane structures is controlled by interactions between specific proteins and lipids. Mitochondrial structure and function depend on regulated fusion and the division of both the outer and inner membranes. Here we discuss recent advances in the regulation of mitochondrial dynamics by two critical phospholipids, phosphatidic acid (PA) and cardiolipin (CL). These two lipids interact with the core components of mitochondrial fusion and division (Opa1, mitofusin, and Drp1) to activate and inhibit these dynamin-related GTPases. Moreover, lipid-modifying enzymes such as phospholipases and lipid phosphatases may organize local lipid composition to spatially and temporarily coordinate a balance between fusion and division to establish mitochondrial morphology.
Product Description
Avanti
TopFluor TMR PA, Avanti Research - A Croda Brand 810240P, powder
Avanti
TopFluor TMR PA, Avanti Research - A Croda Brand 810240C