Merck
  • Beneficial effects of quinoline-3-carboxamide (ABR-215757) on atherosclerotic plaque morphology in S100A12 transgenic ApoE null mice.
Beneficial effects of quinoline-3-carboxamide (ABR-215757) on atherosclerotic plaque morphology in S100A12 transgenic ApoE null mice.
Atherosclerosis (2013-03-19)
Ling Yan, Per Bjork, Radu Butuc, Joseph Gawdzik, Judy Earley, Gene Kim, Marion A Hofmann Bowman
ABSTRACT
There is an emerging widespread interest in the role of damage-associated molecular pattern molecules (DAMP) S100A8, S100A9 and S100A12 in cardiovascular and other diseases. In this study we tested the efficacy of ABR-215757, a S100 protein binding immuno-modulatory compound to stabilize atherosclerosis in transgenic ApoE null mice that express the human pro-inflammatory S100A12 protein within the smooth muscle cell (SM22α-S100A12). Twelve-week old S100A12 transgenic/ApoE(-/-) and WT/ApoE(-/-) mice were treated with ABR-21575 for 5 weeks and were analyzed 4 month later. Surface plasmon resonance analysis demonstrated that S100A12 interacts with ABR-215757 in a zinc dependent manner in vitro. In vivo, ABR-215757 administration reduced features of advanced plaque morphology resulting in smaller necrotic cores, diminished intimal and medial vascular calcification, and reduced amount of infiltrating inflammatory cells. ABR-215757 normalized aortic expression of RAGE protein and normalized experimentally-induced delayed hypersensitivity. The effect of ABR-215757 was more prominent in ApoE(-/-) mice expressing S100A12 than in ApoE(-/-) animals lacking expression of human S100A12 protein. Our data suggest that S100A12 is important for progression of atherosclerosis and can be targeted by the small molecule ABR-215757. The specific binding of quinoline-3-carboxamides to S100A12 attenuates S100A12-mediated features of accelerated murine atherosclerosis.
Sigma-Aldrich
Paquinimod, ≥98% (HPLC)