Merck
Total Synthesis of a Mycolic Acid from Mycobacterium tuberculosis.
Angewandte Chemie (International ed. in English) (2020-02-19)
Nabil Tahiri, Peter Fodran, Dhineshkumar Jayaraman, Jeffrey Buter, Martin D Witte, Tonatiuh A Ocampo, D Branch Moody, Ildiko Van Rhijn, Adriaan J Minnaard
ABSTRACT
In Mycobacterium tuberculosis, mycolic acids and their glycerol, glucose, and trehalose esters ("cord factor") form the main part of the mycomembrane. Despite their first isolation almost a century ago, full stereochemical evaluation is lacking, as is a scalable synthesis required for accurate immunological, including vaccination, studies. Herein, we report an efficient, convergent, gram-scale synthesis of four stereo-isomers of a mycolic acid and its glucose ester. Binding to the antigen presenting protein CD1b and T cell activation studies are used to confirm the antigenicity of the synthetic material. The absolute stereochemistry of the syn-methoxy methyl moiety in natural material is evaluated by comparing its optical rotation with that of synthetic material.
Product Description
Avanti
α-mycolic acid, methoxy cis, powder
Pricing and availability is not currently available.