Merck
  • Lysophosphatidylcholine is generated by spontaneous deacylation of oxidized phospholipids.
Lysophosphatidylcholine is generated by spontaneous deacylation of oxidized phospholipids.
Chemical research in toxicology (2010-10-27)
Jaewoo Choi, Wujuan Zhang, Xiaodong Gu, Xi Chen, Li Hong, James M Laird, Robert G Salomon
ABSTRACT
Elevated levels of lysophosphatidylcholine (lysoPC), present in oxidatively damaged low-density lipoprotein (oxLDL), are implicated in cardiovascular complications. LysoPC is generated by free radical-catalyzed oxidation of polyunsaturated PCs to oxidatively truncated phosphophatidylcholines (oxPCs). It is known that oxPCs are especially susceptible to hydrolysis by platelet-activating factor acetylhydrolase, a phospholipase (PL) A(2) that exists in plasma largely in association with LDL. Drugs that aim to prevent the generation of lysoPC by inhibiting this PLA(2)-catalyzed hydrolysis are in advanced clinical trials. We now report that spontaneous deacylation oxPCs, such as 1-palmityl-2-(4-hydroxy-7-oxo-5-heptenoyl)-sn-glycero-3-phosphocholine, occurs readily under physiological conditions of temperature and pH (t(1/2) = 30 min at 37 °C and pH 7.4). We also show that this reaction proceeds through an intramolecular transesterification mechanism. Because antiphospholipase drugs cannot block this nonenzymatic pathway to lysoPC, additional therapeutic measures may be needed to avoid the pathological consequences of the newly discovered biomolecular chemistry of oxPCs.
Product Description
Avanti
13:0 Lyso LPC, Avanti Research - A Croda Brand