Merck
  • Data Processing Pipeline for Lipid Profiling of Carotid Atherosclerotic Plaque with Mass Spectrometry Imaging.
Data Processing Pipeline for Lipid Profiling of Carotid Atherosclerotic Plaque with Mass Spectrometry Imaging.
Journal of the American Society for Mass Spectrometry (2019-06-30)
Mirjam Visscher, Astrid M Moerman, Peter C Burgers, Heleen M M Van Beusekom, Theo M Luider, Hence J M Verhagen, Antonius F W Van der Steen, Kim Van der Heiden, Gijs Van Soest
ABSTRACT
Atherosclerosis is a lipid and inflammation-driven disease of the arteries that is characterized by gradual buildup of plaques in the vascular wall. A so-called vulnerable plaque, consisting of a lipid-rich necrotic core contained by a thin fibrous cap, may rupture and trigger thrombus formation, which can lead to ischemia in the heart (heart attack) or in the brain (stroke). In this study, we present a protocol to investigate the lipid composition of advanced human carotid plaques using matrix-assisted laser desorption ionization (MALDI) mass spectrometry imaging (MSI), providing a framework that should enable the discrimination of vulnerable from stable plaques based on lipid composition. We optimized the tissue preparation and imaging methods by systematically analyzing data from three specimens: two human carotid endarterectomy samples (advanced plaque) and one autopsy sample (early stage plaque). We show a robust data reduction method and evaluate the variability of the endarterectomy samples. We found diacylglycerols to be more abundant in a thrombotic area compared to other plaque areas and could distinguish advanced plaque from early stage plaque based on cholesteryl ester composition. We plan to use this systematic approach to analyze a larger dataset of carotid atherosclerotic plaques.
Product Description
Avanti
17:0 cholesteryl ester, cholest-5-en-3β-yl heptadecanoate, methylene chloride solution
Pricing and availability is not currently available.
Sigma-Aldrich
Iron(III) chloride, reagent grade, 97%
Pricing and availability is not currently available.