Merck
  • Streptococcus pneumoniae Infection Promotes Histone H3 Dephosphorylation by Modulating Host PP1 Phosphatase.
Streptococcus pneumoniae Infection Promotes Histone H3 Dephosphorylation by Modulating Host PP1 Phosphatase.
Cell reports (2020-03-27)
Wenyang Dong, Orhan Rasid, Christine Chevalier, Michael Connor, Matthew J G Eldridge, Melanie Anne Hamon
ABSTRACT
Pathogenic bacteria can alter host gene expression through post-translational modifications of histones. We show that a natural colonizer, Streptococcus pneumoniae, induces specific histone modifications, including robust dephosphorylation of histone H3 on serine 10 (H3S10), during infection of respiratory epithelial cells. The bacterial pore-forming toxin pneumolysin (PLY), along with the pyruvate oxidase SpxB responsible for H2O2 production, play important roles in the induction of this modification. The combined effects of PLY and H2O2 trigger host signaling that culminates in H3S10 dephosphorylation, which is mediated by the host cell phosphatase PP1. Strikingly, S. pneumoniae infection induces dephosphorylation and subsequent activation of PP1 catalytic activity. Colonization of PP1 catalytically deficient cells results in impaired intracellular S. pneumoniae survival and infection. Interestingly, PP1 activation and H3S10 dephosphorylation are not restricted to S. pneumoniae and appear to be general epigenomic mechanisms favoring intracellular survival of pathogenic bacteria.
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid
Sigma-Aldrich
Nicotinamide, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
PP1, ≥98% (HPLC)
Sigma-Aldrich
Anti-PP1β Antibody, Upstate®, from rabbit