Merck
  • Insight into Hyper-Branched Aluminum Phosphonate in Combination with Multiple Phosphorus Synergies for Fire-Safe Epoxy Resin Composites.
Insight into Hyper-Branched Aluminum Phosphonate in Combination with Multiple Phosphorus Synergies for Fire-Safe Epoxy Resin Composites.
Polymers (2020-01-08)
Yao Yuan, Bin Yu, Yongqian Shi, Long Mao, Jianda Xie, Haifeng Pan, Yuejun Liu, Wei Wang
ABSTRACT
Epoxy resin (EP) has widespread applications in thermosetting materials with great versatility and desirable properties such as high electrical resistivity and satisfactory mechanical properties. At present, 9,10-Dihydro-9-oxa-10-phosphaphenanthrene-10-oxide (DOPO) is widely applied to EP matrix for high flame resistance. Nevertheless, EP/DOPO composites acquire highly toxic decomposition products and smoke particles produced during combustion due to the gaseous fire-inhibition mechanism, which will be a major problem. To address this concern, an effective hyper-branched aluminum phosphonate (AHPP) was rationally designed and then coupled with DOPO into EP matrix to fabricate the fire-safe epoxy resin composites. On the basis of the results, significant increment in limiting oxygen index value (an achievement of 32% from 23.5% for pristine EP) and reduction in peak heat release rate and total heat release (59.4% and 45.6%) with the DOPO/AHPP ratio of 2:1 were recorded. During the cone calorimeter test, both the smoke production and total CO yield of EP-4 composite with the DOPO/AHPP ratio of 1:2 were dramatically decreased by 42.7% and 53.6%, which was mainly associated with the excellent catalytic carbonization of AHPP submicro-particles for EP composite. Future applications of submicro-scaled flame-retardant with various phosphorus oxidation states will have good prospects for development.
Sigma-Aldrich
Bis[tetrakis(hydroxymethyl)phosphonium] sulfate solution, technical, 70-75% in H2O (T)
Pricing and availability is not currently available.