Merck
  • Hotspots of Aberrant Enhancer Activity in Fibrolamellar Carcinoma Reveal Candidate Oncogenic Pathways and Therapeutic Vulnerabilities.
Hotspots of Aberrant Enhancer Activity in Fibrolamellar Carcinoma Reveal Candidate Oncogenic Pathways and Therapeutic Vulnerabilities.
Cell reports (2020-04-16)
Timothy A Dinh, Ramja Sritharan, F Donelson Smith, Adam B Francisco, Rosanna K Ma, Rodica P Bunaciu, Matt Kanke, Charles G Danko, Andrew P Massa, John D Scott, Praveen Sethupathy
ABSTRACT
Fibrolamellar carcinoma (FLC) is a rare, therapeutically intractable liver cancer that disproportionately affects youth. Although FLC tumors exhibit a distinct gene expression profile, the chromatin regulatory landscape and the genes most critical for tumor cell survival remain unclear. Here, we use chromatin run-on sequencing to discover ∼7,000 enhancers and 141 enhancer hotspots activated in FLC relative to nonmalignant liver. Bioinformatic analyses reveal aberrant ERK/MEK signaling and candidate master transcriptional regulators. We also define the genes most strongly associated with hotspots of FLC enhancer activity, including CA12 and SLC16A14. Treatment of FLC cell models with inhibitors of CA12 or SLC16A14 independently reduce cell viability and/or significantly enhance the effect of the MEK inhibitor cobimetinib. These findings highlight molecular targets for drug development, as well as drug combination approaches.
Sigma-Aldrich
U-104, ≥98% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
L-(−)-Glucose, ≥99%
Pricing and availability is not currently available.