- Rapid access to 1,6-anhydro-beta-L-hexopyranose derivatives via domino reaction: synthesis of L-allose and L-glucose.
Rapid access to 1,6-anhydro-beta-L-hexopyranose derivatives via domino reaction: synthesis of L-allose and L-glucose.
An expeditious and efficient synthesis of 1,6-anhydro-beta-L-hexopyranosyl derivatives 3 as valuable building blocks for the preparation of L-sugars is herein reported. This route relies upon the use of a domino reaction involving five synthetic steps from the 5,6-dihydro-1,4-dithiin 4. As 1,6-anhydro derivatives 3 are obtained, dithioethylene bridge removal and double-bond dihydroxylation give access to protected L-allose and L-glucose in stereoselective fashion and high yields.