Merck
  • A general approach for hysteresis-free, operationally stable metal halide perovskite field-effect transistors.
A general approach for hysteresis-free, operationally stable metal halide perovskite field-effect transistors.
Science advances (2020-04-18)
Satyaprasad P Senanayak, Mojtaba Abdi-Jalebi, Varun S Kamboj, Remington Carey, Ravichandran Shivanna, Tian Tian, Guillaume Schweicher, Junzhan Wang, Nadja Giesbrecht, Daniele Di Nuzzo, Harvey E Beere, Pablo Docampo, David A Ritchie, David Fairen-Jimenez, Richard H Friend, Henning Sirringhaus
ABSTRACT
Despite sustained research, application of lead halide perovskites in field-effect transistors (FETs) has substantial concerns in terms of operational instabilities and hysteresis effects which are linked to its ionic nature. Here, we investigate the mechanism behind these instabilities and demonstrate an effective route to suppress them to realize high-performance perovskite FETs with low hysteresis, high threshold voltage stability (ΔVt < 2 V over 10 hours of continuous operation), and high mobility values >1 cm2/V·s at room temperature. We show that multiple cation incorporation using strain-relieving cations like Cs and cations such as Rb, which act as passivation/crystallization modifying agents, is an effective strategy for reducing vacancy concentration and ion migration in perovskite FETs. Furthermore, we demonstrate that treatment of perovskite films with positive azeotrope solvents that act as Lewis bases (acids) enables a further reduction in defect density and substantial improvement in performance and stability of n-type (p-type) perovskite devices.
Sigma-Aldrich
Tin(IV) iodide, anhydrous, powder, 99.999% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Rubidium iodide, 99.9% trace metals basis
Pricing and availability is not currently available.