Merck
  • Properties of Rigid Polyurethane Foam Modified by Tung Oil-Based Polyol and Flame-Retardant Particles.
Properties of Rigid Polyurethane Foam Modified by Tung Oil-Based Polyol and Flame-Retardant Particles.
Polymers (2020-01-18)
Wei Zhou, Shu-Jie Hao, Guo-Dong Feng, Pu-You Jia, Xiao-Li Ren, Meng Zhang, Yong-Hong Zhou
ABSTRACT
Although tung oil is renewable, with an abundant production and low price in China, and it is used to synthesize different polyols for rigid polyurethane foam (RPUF), it remains a challenge to improve the properties of RPUF by redesigning the formula. Therefore, we propose four novel compounds to strengthen the properties of RPUF, such as the catalyst-free synthesis of tung oil-based polyol (PTOK), aluminum phosphate micro-capsule (AM), silica micro-capsule (SiM), and grafted epoxidized monoglyceride of tung oil on the surface of SiO2 (SiE), which were designed and introduced into the RPUF. Because of the PTOK with a catalytic function, the foaming process of some RPUF samples was catalyst-free. The results show that the incorporation of AM, SiM, and SiE, respectively, endow RPUF with a better thermal stability at a high temperature, and the T5%, Tmax1, and Tmax2 of RPUF appeared to be reduced, however, the Tmax3 and residue rate at 800 °C were improved, which may have a positive effect on the extension of the rescue time in case of fire, and the limiting oxygen index (LOI) value was increased to 22.6%. The formula, containing 25% PTOK made the RPUF environment-friendly. The results were obtained by comparing the pore size and mechanical properties of the RPUF-the AM had a better dispersion in the foam, and the foam obtained a better mechanical, thermal, and flame retardancy.
Sigma-Aldrich
Edelfosine, ≥95% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
3-(2-Aminoethylamino)propyldimethoxymethylsilane, ≥95.0%
Pricing and availability is not currently available.