Merck
  • Multivariate Analysis for Molecular Species of Cholesteryl Ester in the Human Serum.
Multivariate Analysis for Molecular Species of Cholesteryl Ester in the Human Serum.
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry (2019-11-19)
Yifan Chen, Shu-Ping Hui, Yusuke Miura, Sota Kato, Toshihiro Sakurai, Zhen Chen, Emiko Okada, Shigekazu Ukawa, Takafumi Nakagawa, Koshi Nakamura, Akiko Tamakoshi, Hitoshi Chiba, Hiroyuki Minami, Masahiro Mizuta
ABSTRACT
Cholesteryl ester (CE) is an ester of cholesterol and fatty acid (FA). Plasma CE reflects complicated metabolisms of cholesterol, phospholipids, lipoproteins, and dietary FAs. An informatics approach could be useful for analysis of the CE species. In this study, two basic dimension reduction methods, principal component analysis (PCA) and factor analysis, were applied to serum CE species determined by LC-MS/MS in a Japanese population (n = 545). PCA and factor analysis both reflected the size (concentration), food source, fat solubility, and biological aspect of the CE species. In a comparison between PCA (PC4) and factor analysis (factor 4), the latter was found to be more suggestive from a biological aspect of n-6 FAs. Cholesteryl docosahexaenoate (DHA) was found to be unique by a factor analysis, possibly relevant to the unique accumulation of DHA in the brain. An informatics approach, especially factor analysis, might be useful for the analysis of complicated metabolism of CE species in the serum.
Sigma-Aldrich
N-(3-Dimethylaminopropyl)-N′-ethylcarbodiimide, ≥97.0% (T)
Pricing and availability is not currently available.
Supelco
Fructose Assay Kit, sufficient for 20 assays
Pricing and availability is not currently available.