Merck
  • In vivo sodium concentration continuously monitored with fluorescent sensors.
In vivo sodium concentration continuously monitored with fluorescent sensors.
Integrative biology : quantitative biosciences from nano to macro (2010-11-23)
J Matthew Dubach, Edward Lim, Ning Zhang, Kevin P Francis, Heather Clark
ABSTRACT
Sodium balance is vital to maintaining normal physiological function. Imbalances can occur in a variety of diseases, during certain surgical operations or during rigorous exercise. There is currently no method to continuously monitor sodium concentration in patients who may be susceptible to hyponatremia. Our approach was to design sodium specific fluorescent sensors capable of measuring physiological fluctuations in sodium concentration. The sensors are submicron plasticized polymer particles containing sodium recognition components that are coated with biocompatible poly(ethylene) glycol. Here, the sensors were brought up in saline and placed in the subcutaneous area of the skin of mice by simple injection. The fluorescence was monitored in real time using a whole animal imager to track changes in sodium concentrations. This technology could be used to monitor certain disease states or warn against dangerously low levels of sodium during exercise.
Product Description
Avanti
18:0 PEG550 PE, Avanti Research - A Croda Brand 880520P, powder
Pricing and availability is not currently available.