MilliporeSigma
  • Effects of cholesterol on phospholipid membranes: inhibition of the interdigitated gel phase of F-DPPC and F-DPPC/DPPC.
Effects of cholesterol on phospholipid membranes: inhibition of the interdigitated gel phase of F-DPPC and F-DPPC/DPPC.
Chemistry and physics of lipids (2011-12-28)
Eric A Smith, Weidong Wang, Phoebe K Dea
ABSTRACT
Unlike the parent phospholipid, 1,2-dipalmitoyl-sn-glycero-3-phosphocholine (DPPC), the monofluorinated analog, 1-palmitoyl-2-(16-fluoropalmitoyl)sn-glycero-3-phosphocholine (F-DPPC), spontaneously forms an interdigitated gel phase (L(β)I) below the main transition temperature (T(m)). We have examined the effects of introducing cholesterol to F-DPPC and 1:1 F-DPPC/DPPC membranes using a combination of DSC, optical density, fluorescence intensity and polarization, (31)P NMR, and X-ray diffraction techniques. Cholesterol increases the fluidity of the gel phase, broadens the main transition, and decreases the main transition enthalpy. However, these results also reveal that there is an unusually large degree of phase coexistence between the L(β)I and non-interdigitated gel phases when cholesterol is added. Cholesterol encourages this phase segregation by partitioning into the thicker non-interdigitated domains. At higher cholesterol concentrations, the majority or all of the L(β)I phase of F-DPPC and 1:1 F-DPPC/DPPC is eliminated and is replaced by a non-interdigitated liquid-ordered (l(o)) phase with properties similar to DPPC/cholesterol. Consequently, cholesterol mitigates the influence the CF moiety has on the thermodynamic phase behavior of F-DPPC. Our findings demonstrate that there are multiple characteristics of cholesterol-rich membranes that disfavor interdigitation.