Merck
  • Correlation Study of Honey Regarding their Physicochemical Properties and Sugars and Cyclitols Content.
Correlation Study of Honey Regarding their Physicochemical Properties and Sugars and Cyclitols Content.
Molecules (Basel, Switzerland) (2019-12-22)
Ileana Andreea Ratiu, Hossam Al-Suod, Małgorzata Bukowska, Magdalena Ligor, Bogusław Buszewski
ABSTRACT
Honey is a natural sweetener, with an osmotic effect on microorganisms due to the increased sugar content and low amount of water. Cyclitols are minor constituents of honey. They play a defensive role in plants against unfavorable environmental conditions. Honey's physicochemical properties can vary, resulting in a wide range of colors, flavors, scents, antioxidant activity, dissimilar values of pH, acidity, electrical conductivity, etc. Some literature regarding correlation between honey types is already available, but a comprehensive study displaying an ample evaluation of multifarious aspects is still needed. This study focuses on the correlation between 18 honey types, originating from 10 countries, collected during four years, summarizing a total of 38 samples. A total of 6 physicochemical properties and 18 target components (sugars and cyclitols) were considered as variables. A correlation analysis is presented between the investigated parameters and between honey types, together with the statistical analysis which allowed for observation of the clusters' distribution according with the investigated variables.
Sigma-Aldrich
allo-Inositol, 97%
Pricing and availability is not currently available.