Merck
  • Silencing of glycopeptide resistance in Enterococcus faecalis BM4405 by novobiocin.
Silencing of glycopeptide resistance in Enterococcus faecalis BM4405 by novobiocin.
Antimicrobial agents and chemotherapy (2005-03-29)
Lorena Abadía Patiño, Marc Chippaux, Patrice Courvalin, Bruno Périchon
ABSTRACT
Enterococcus faecalis BM4405-1, a susceptible derivative of the VanE-type vancomycin-resistant E. faecalis strain BM4405, was obtained after growth in the presence of novobiocin, an inhibitor of the GyrB subunit of DNA gyrase. In contrast to findings for BM4405, UDP-MurNAc-L-Ala-gamma-D-Glu-L-Lys-D-Ala-D-Ala (pentapeptide[D-Ala]) was the only peptidoglycan precursor found in BM4405-1, and no VanXY(E) D,D-peptidase or VanT serine racemase activities were detected in that strain, even after induction by subinhibitory concentrations of vancomycin. Sequencing of the vanE operon of BM4405-1 revealed two mutations leading to substitutions in VanE (D200N) and in the C-terminal amino acid of VanR(E) (Y225F). Cloning of the vanE, vanXY(E), and vanT(E) genes of BM4405-1 into the susceptible E. faecalis strain JH2-2 conferred resistance to vancomycin, indicating that the mutation in vanE was not responsible for susceptibility. Transcriptional analysis of the vanE operon in BM4405 by quantitative reverse transcription-PCR indicated that novobiocin did not affect the expression level of the vanE operon. Sequencing of the gyrB gene of BM4405-1 revealed a mutation responsible for substitution of a residue (K337Y) required for ATPase activity and thus implicated in DNA supercoiling. Cloning of the gyrB gene of BM4405 restored vancomycin resistance to BM4405-1. Taken together, these data suggest that alteration of DNA supercoiling following a mutation in GyrB was responsible for lack of expression of the vanE operon and thus for vancomycin susceptibility in BM4405-1.
Sigma-Aldrich
Ala-D-γ-Glu-Lys-D-Ala-D-Ala, ≥97% (HPLC)
Pricing and availability is not currently available.