Merck
  • Chromogranin A proteolysis to generate beta-granin and WE-14 in the adenohypophysis during the rat oestrous cycle.
Chromogranin A proteolysis to generate beta-granin and WE-14 in the adenohypophysis during the rat oestrous cycle.
Regulatory peptides (2003-07-23)
Carmel Mary McVicar, Rodat Therese Cunningham, Neil McClure, William James Curry
ABSTRACT
Immunohistochemical analysis of the male and female rat adenohypophysis revealed that chromogranin A (CgA), beta-granin and WE-14 immunostaining was localised to follicle stimulating hormone (FSH) producing cells, while luteinizing hormone (LH) producing cells exhibited chromogranin A and beta-granin immunostaining. The intensity of chromogranin A, beta-granin and WE-14 immunostaining exhibited variation during the oestrous cycle; weak immunostaining was observed during proestrous and oestrous, corresponding with the lowest cellular concentration of luteinizing and follicle stimulating hormone. Chromogranin A and beta-granin immunostaining were similar in both the male and female (at dioestrous), however, a larger number of more intense WE-14 immunopositive cells were evident in the male adenohypophysis relative to the female at any stage of the cycle. The tissue and plasma concentrations of beta-granin and WE-14 immunoreactivity fluctuated throughout the oestrous cycle. Maximum and minimum beta-granin and WE-14 tissue concentration counterpoised the latent maximum and minimum plasma concentration. Chromatographic analysis of adenohypophysis extracts revealed the degree of chromogranin A proteolysis throughout the oestrous cycle; in contrast, plasma profiles consistently possessed a large chromogranin A-like immunoreactant. This data suggests that chromogranin A biosynthesis, proteolysis and the secretion of its derived peptides parallels that of the gonadotroph hormones throughout the oestrous cycle.