Merck
  • Establishing a Field-Effect Transistor Sensor for the Detection of Mutations in the Tumour Protein 53 Gene (TP53)-An Electrochemical Optimisation Approach.
Establishing a Field-Effect Transistor Sensor for the Detection of Mutations in the Tumour Protein 53 Gene (TP53)-An Electrochemical Optimisation Approach.
Biosensors (2019-12-11)
Lisa Crossley, Bukola Attoye, Vincent Vezza, Ewen Blair, Damion K Corrigan, Stuart Hannah
ABSTRACT
We present a low-cost, sensitive and specific DNA field-effect transistor sensor for the rapid detection of a common mutation to the tumour protein 53 gene (TP53). The sensor consists of a commercially available, low-cost, field-effect transistor attached in series to a gold electrode sensing pad for DNA hybridisation. The sensor has been predominantly optimised electrochemically, particularly with respect to open-circuit potentiometry as a route towards understanding potential (voltage) changes upon DNA hybridisation using a transistor. The developed sensor responds sensitively to TP53 mutant DNA as low as 100 nM concentration. The sensor responds linearly as a function of DNA target concentration and is able to differentiate between complementary and noncomplementary DNA target sequences.
Sigma-Aldrich
3-Mercapto-1-propanol, 95%
Pricing and availability is not currently available.