Merck
  • Programing Assembling/Releasing Multifunctional miRNA Nanomedicine to Treat Prostate Cancer.
Programing Assembling/Releasing Multifunctional miRNA Nanomedicine to Treat Prostate Cancer.
ACS applied materials & interfaces (2020-01-28)
Ding Ma, Hongmei Liu, Peipei Zhao, Li Ye, Hanbing Zou, Xue Zhao, Huili Dai, Xianming Kong, Peifeng Liu
ABSTRACT
MicroRNAs (miRNAs) therapy has shown to have great promise for the treatment of androgen-independent prostate cancer (AIPC) due to the low efficiency of hormonal therapy. However, instability of RNA and inefficiency of RNA therapy limit the use of miRNAs in the treatment of AIPC. Here, we report a pH/ATP-activated nanocomplexes for increasing cytosolic delivery of miR146a which can effectively inhibit the expression of epidermal growth factor receptor (EGFR) in AIPC. The nanocomplexes show identical suppressing effect in invasion, colony formation, migration ability, and growth of DU145 cells compared with Lipofectamine 2000 (lipo). But for in vivo experiments, the nanocomplexes vigorously suppress the growth of tumor volumes comparing to lipo group after five weeks' treatment. These results demonstrate the potential of the pH/ATP-activated nanocarriers for AIPC gene therapy.
Sigma-Aldrich
4-(Bromomethyl)phenylboronic acid
Pricing and availability is not currently available.