Merck
  • BCL-XL directly modulates RAS signalling to favour cancer cell stemness.
BCL-XL directly modulates RAS signalling to favour cancer cell stemness.
Nature communications (2017-10-27)
Sophie de Carné Trécesson, Frédérique Souazé, Agnès Basseville, Anne-Charlotte Bernard, Jessie Pécot, Jonathan Lopez, Margaux Bessou, Kristopher A Sarosiek, Anthony Letai, Sophie Barillé-Nion, Isabelle Valo, Olivier Coqueret, Catherine Guette, Mario Campone, Fabien Gautier, Philippe Paul Juin
ABSTRACT
In tumours, accumulation of chemoresistant cells that express high levels of anti-apoptotic proteins such as BCL-XL is thought to result from the counter selection of sensitive, low expresser clones during progression and/or initial treatment. We herein show that BCL-XL expression is selectively advantageous to cancer cell populations even in the absence of pro-apoptotic pressure. In transformed human mammary epithelial cells BCL-XL favours full activation of signalling downstream of constitutively active RAS with which it interacts in a BH4-dependent manner. Comparative proteomic analysis and functional assays indicate that this is critical for RAS-induced expression of stemness regulators and maintenance of a cancer initiating cell (CIC) phenotype. Resistant cancer cells thus arise from a positive selection driven by BCL-XL modulation of RAS-induced self-renewal, and during which apoptotic resistance is not necessarily the directly selected trait.
Sigma-Aldrich
Anti-Bim Antibody, internal epitope, pan-Bim isoforms, Chemicon®, from rabbit
Sigma-Aldrich
Anti-Actin Antibody, clone C4, clone C4, Chemicon®, from mouse