Merck
  • GAT-1 regulates both tonic and phasic GABA(A) receptor-mediated inhibition in the cerebral cortex.
GAT-1 regulates both tonic and phasic GABA(A) receptor-mediated inhibition in the cerebral cortex.
Journal of neurochemistry (2008-02-06)
Luca Bragina, Ivan Marchionni, Azar Omrani, Andrea Cozzi, Domenico E Pellegrini-Giampietro, Enrico Cherubini, Fiorenzo Conti
ABSTRACT
gamma-Aminobutyric acid 1 (GAT-1) is the most copiously expressed GABA transporter; we studied its role in phasic and tonic inhibition in the neocortex using GAT-1 knockout (KO) mice. Immunoblotting and immunocytochemical studies showed that GAT-2 and GAT-3 levels in KOs were unchanged and that GAT-3 was not redistributed in KOs. Moreover, the expression of GAD65/67 was increased, whereas that of GABA or VGAT was unchanged. Microdialysis studies showed that in KOs spontaneous extracellular release of GABA and glutamate was comparable in WT and KO mice, whereas KCl-evoked output of GABA, but not of glutamate, was significantly increased in KOs. Recordings from layer II/III pyramids revealed a significant increase in GABA(A)R-mediated tonic conductance in KO mice. The frequency, amplitude and kinetics of spontaneous inhibitory post-synaptic currents (IPSCs) were unchanged, whereas the decay time of evoked IPSCs was significantly prolonged in KO mice. In KO mice, high frequency stimulation of GABAergic terminals induced large GABA(A)R-mediated inward currents associated with a reduction in amplitude and decay time of IPSCs evoked immediately after the train. The recovery process was slower in KO than in WT mice. These studies show that in the cerebral cortex of GAT-1 KO mice GAT-3 is not redistributed and GADs are adaptively changed and indicate that GAT-1 has a prominent role in both tonic and phasic GABA(A)R-mediated inhibition, in particular during sustained neuronal activity.
Sigma-Aldrich
Anti-Glutamate Decarboxylase 65 & 67 Antibody, Chemicon®, from rabbit
Sigma-Aldrich
Anti-GABA Antibody, serum, Chemicon®