Merck
  • Leptin enhances the recruitment of endothelial progenitor cells into neointimal lesions after vascular injury by promoting integrin-mediated adhesion.
Leptin enhances the recruitment of endothelial progenitor cells into neointimal lesions after vascular injury by promoting integrin-mediated adhesion.
Circulation research (2008-07-29)
Marco R Schroeter, Maren Leifheit, Philipp Sudholt, Nana-Maria Heida, Claudia Dellas, Ilonka Rohm, Frauke Alves, Marta Zientkowska, Stavros Rafail, Miriam Puls, Gerd Hasenfuss, Stavros Konstantinides, Katrin Schäfer
ABSTRACT
The adipocytokine leptin modulates vascular remodeling and neointima formation. Because endothelial progenitor cells (EPCs) participate in vascular repair, we analyzed the effects of leptin on human EPC function in vitro and in vivo. After 7 days in culture, EPCs expressed the leptin receptor and responded to leptin stimulation with increased STAT3 phosphorylation. Incubation of EPCs with leptin (at concentrations between 1 and 100 ng/mL) increased the number of EPCs adhering to vitronectin and fibronectin in a receptor-specific manner. It also enhanced the capacity of EPCs to incorporate into a monolayer of human endothelial cells and the adherence of these cells to activated platelets. Leptin upregulated alphavbeta5 and alpha4 integrin expression in EPCs, and the effects of leptin on EPC function could be prevented, at least in part, by RGD peptides and function-blocking antibodies. Intravenous injection of fluorescently labeled human EPCs into athymic nude mice shortly after vascular injury revealed that preincubation of EPCs with leptin augmented their accumulation within intimal lesions, accelerating reendothelialization and decreasing neointima formation in an alphavbeta5 and alpha4 integrin-dependent manner. Our findings suggest that leptin specifically modulates the adhesive properties and the homing potential of EPCs and may thus enhance their capacity to promote vascular regeneration in vivo.
Sigma-Aldrich
Anti-Integrin αVβ5 Antibody, clone P1F6, clone P1F6, Chemicon®, from mouse
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-Integrin α4 Antibody, clone P1H4, clone P1H4, Chemicon®, from mouse
Pricing and availability is not currently available.