Merck
  • Differential expression of GAP-43 and neurofilament during peripheral nerve regeneration through bio-artificial conduits.
Differential expression of GAP-43 and neurofilament during peripheral nerve regeneration through bio-artificial conduits.
Journal of tissue engineering and regenerative medicine (2014-08-02)
Víctor Carriel, Ingrid Garzón, Antonio Campos, Maria Cornelissen, Miguel Alaminos
ABSTRACT
Nerve conduits are promising alternatives for repairing nerve gaps; they provide a close microenvironment that supports nerve regeneration. In this sense, histological analysis of axonal growth is a determinant to achieve successful nerve regeneration. To evaluate this process, the most-used immunohistochemical markers are neurofilament (NF), β-III tubulin and, infrequently, GAP-43. However, GAP-43 expression in long-term nerve regeneration models is still poorly understood. In this study we analysed GAP-43 expression and its correlation with NF and S-100, using three tissue-engineering approaches with different regeneration profiles. A 10 mm gap was created in the sciatic nerve of 12 rats and repaired using collagen conduits or collagen conduits filled with fibrin-agarose hydrogels or with hydrogels containing autologous adipose-derived mesenchymal stem cells (ADMSCs). After 12 weeks the conduits were harvested for histological analysis. Our results confirm the long-term expression of GAP-43 in all groups. The expression of GAP-43 and NF was significantly higher in the group with ADMSCs. Interestingly, GAP-43 was observed in immature, newly formed axons and NF in thicker and mature axons. These proteins were not co-expressed, demonstrating their differential expression in newly formed nerve fascicles. Our descriptive and quantitative histological analysis of GAP-43 and NFL allowed us to determine, with high accuracy, the heterogenic population of axons at different stages of maturation in three tissue-engineering approaches. Finally, to perform a complete assessment of axonal regeneration, the quantitative immunohistochemical evaluation of both GAP-43 and NF could be a useful quality control in tissue engineering. Copyright © 2014 John Wiley & Sons, Ltd.
Sigma-Aldrich
Anti-GAP43 (Ab-41) antibody produced in rabbit, affinity isolated antibody
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-Mouse IgG (whole molecule)–FITC antibody produced in goat, affinity isolated antibody, buffered aqueous solution
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-Neurofilament 160/200 antibody, Mouse monoclonal, ~2 mg/mL, clone RMdO20, purified from hybridoma cell culture
Pricing and availability is not currently available.