Merck
  • Synergistic action of nectins and cadherins generates the mosaic cellular pattern of the olfactory epithelium.
Synergistic action of nectins and cadherins generates the mosaic cellular pattern of the olfactory epithelium.
The Journal of cell biology (2016-03-02)
Sayaka Katsunuma, Hisao Honda, Tomoyasu Shinoda, Yukitaka Ishimoto, Takaki Miyata, Hiroshi Kiyonari, Takaya Abe, Ken-Ichi Nibu, Yoshimi Takai, Hideru Togashi
ABSTRACT
In the olfactory epithelium (OE), olfactory cells (OCs) and supporting cells (SCs), which express different cadherins, are arranged in a characteristic mosaic pattern in which OCs are enclosed by SCs. However, the mechanism underlying this cellular patterning is unclear. Here, we show that the cellular pattern of the OE is established by cellular rearrangements during development. In the OE, OCs express nectin-2 and N-cadherin, and SCs express nectin-2, nectin-3, E-cadherin, and N-cadherin. Heterophilic trans-interaction between nectin-2 on OCs and nectin-3 on SCs preferentially recruits cadherin via α-catenin to heterotypic junctions, and the differential distributions of cadherins between junctions promote cellular intercalations, resulting in the formation of the mosaic pattern. These observations are confirmed by model cell systems, and various cellular patterns are generated by the combinatorial expression of nectins and cadherins. Collectively, the synergistic action of nectins and cadherins generates mosaic pattern, which cannot be achieved by a single mechanism.
Sigma-Aldrich
Anti-β-Catenin Antibody, clone 5H10, clone 5H10, Chemicon®, from mouse
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-ZO-1 Antibody, clone R40.76, clone R40.76, from rat
Pricing and availability is not currently available.