Merck
  • Neonatal maternal deprivation impairs localized de novo activity-induced protein translation at the synapse in the rat hippocampus.
Neonatal maternal deprivation impairs localized de novo activity-induced protein translation at the synapse in the rat hippocampus.
Bioscience reports (2018-04-28)
Faraz Ahmad, Mohammad Salahuddin, Khaldoon Alsamman, Hatem K Herzallah, Sultan T Al-Otaibi
ABSTRACT
Neonatal neuropsychiatric stress induces alterations in neurodevelopment that can lead to irreversible damage to neuronal physiology, and social, behavioral, and cognitive skills. In addition, this culminates to an elevated vulnerability to stress and anxiety later in life. Developmental deficits in hippocampal synaptic function and plasticity are among the primary contributors of detrimental alterations in brain function induced by early-life stress. However, the underlying molecular mechanisms are not completely understood. Localized protein translation, occurring at the synapse and triggered by neuronal activity, is critical for synapse function, maintenance, and plasticity. We used a rodent model of chronic maternal deprivation to characterize the effects of early-life neuropsychiatric stress on localized de novo protein translation at synaptic connections between neurons. Synaptoneurosomal preparations isolated biochemically from the hippocampi of rat pups that were subjected to maternal deprivation were deficient in depolarization-induced activity-dependent protein translation when compared with littermate controls. Conversely, basal unstimulated protein translation was not affected. Moreover, deficits in activity-driven synaptic protein translation were significantly correlated with a reduction in phosphorylated cell survival protein kinase protein B or Akt (p473 Ser and p308 Thr), but not phosphorylated extracellular signal-regulated kinase.
Sigma-Aldrich
Phosphatase Inhibitor Cocktail 2, aqueous solution (dark coloration may develop upon storage, which does not affect the activity)
Sigma-Aldrich
Anti-Puromycin Antibody, clone 17H1, clone 17H1, from rat