Merck
  • Intestinal calcium absorption in rats is stimulated by dietary lactulose and other resistant sugars.
Intestinal calcium absorption in rats is stimulated by dietary lactulose and other resistant sugars.
The Journal of nutrition (1993-12-01)
R Brommage, C Binacua, S Antille, A L Carrié
ABSTRACT
Lactulose is a disaccharide analogue of lactose that is resistant to metabolism in the small intestine but not in the large intestine. The effects of lactulose and other sugars on intestinal Ca absorption were determined from the decrease in the 47Ca:47 Sc ratio between diet and feces after feeding male rats diets containing these sugars during a single night. Dietary lactulose was more potent than lactose in stimulating Ca absorption and was effective between 5 and 38 wk of age. The component sugars of lactulose, galactose and fructose, did not influence Ca absorption when provided together at concentrations equimolar to that of lactulose. The stimulation of Ca absorption by dietary lactulose increased as dietary Ca concentration was raised and was not influenced by prior injections of calcitriol. Lactulose must be present in the same meal as Ca to stimulate Ca absorption, but this stimulation was lost if the rats were fed lactulose continuously for 2 or 7 d prior to the test diet. Other sugars thought to be poorly absorbed in the small intestine (xylitol, lactobionate, arabinose, raffinose, pyroglutamate, sorbitol, gluconate and raftilose) stimulated Ca absorption to an identical extent as lactulose. Cecectomy did not influence the enhancement of Ca absorption by lactulose. These results indicate that sugars resistant to metabolism and absorption in the small intestine but not the large intestine stimulate Ca absorption in the small intestine.
Sigma-Aldrich
Lactobionic acid, ≥97% (TLC), cell impermeant agent