Merck
  • Surface LAMP-2 Is an Endocytic Receptor That Diverts Antigen Internalized by Human Dendritic Cells into Highly Immunogenic Exosomes.
Surface LAMP-2 Is an Endocytic Receptor That Diverts Antigen Internalized by Human Dendritic Cells into Highly Immunogenic Exosomes.
Journal of immunology (Baltimore, Md. : 1950) (2017-06-14)
Dario Armando Leone, Andrea Peschel, Markus Brown, Helga Schachner, Miriam J Ball, Marianna Gyuraszova, Ulrike Salzer-Muhar, Minoru Fukuda, Caterina Vizzardelli, Barbara Bohle, Andrew J Rees, Renate Kain
ABSTRACT
The lysosome-associated membrane protein (LAMP) family includes the dendritic cell endocytic receptors DC-LAMP and CD68, as well as LAMP-1 and LAMP-2. In this study we identify LAMP-1 (CD107a) and LAMP-2 (CD107b) on the surface of human monocyte-derived dendritic cells (MoDC) and show only LAMP-2 is internalized after ligation by specific Abs, including H4B4, and traffics rapidly but transiently to the MHC class II loading compartment, as does Ag conjugated to H4B4. However, pulsing MoDC with conjugates of primary (keyhole limpet hemocyanin; KLH) and recall (Bet v 1) Ags (H4B4*KLH and H4B4*Bet v 1) induced significantly less CD4 cell proliferation than pulsing with native Ag or Ag conjugated to control mAb (ISO*KLH and ISO*Bet v 1). In H4B4*KLH-pulsed MoDC, the duration of KLH residence in MHC class II loading compartments was significantly reduced, as were surface HLA-DR and DR-bound KLH-derived peptides. Paradoxically, MoDC pulsed with H4B4*KLH, but not the other KLH preparations, induced robust proliferation of CD4 cells separated from them by a transwell membrane, indicating factors in the supernatant were responsible. Furthermore, extracellular vesicles from supernatants of H4B4*KLH-pulsed MoDC contained significantly more HLA-DR and KLH than those purified from control MoDC, and KLH was concentrated specifically in exosomes that were a uniquely effective source of Ag in standard T cell proliferation assays. In summary, we identify LAMP-2 as an endocytic receptor on human MoDC that routes cargo into unusual Ag processing pathways, which reduces surface expression of Ag-derived peptides while selectively enriching Ag within immunogenic exosomes. This novel pathway has implications for the initiation of immune responses both locally and at distant sites.
Sigma-Aldrich
Anti-Hemocyanin (KLH) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Pricing and availability is not currently available.