Merck
  • Gene expression in proliferating cells of the dinoflagellate Alexandrium catenella (Dinophyceae).
Gene expression in proliferating cells of the dinoflagellate Alexandrium catenella (Dinophyceae).
Applied and environmental microbiology (2010-05-04)
Eve Toulza, Mi-Sun Shin, Guillaume Blanc, Stéphane Audic, Mohamed Laabir, Yves Collos, Jean-Michel Claverie, Daniel Grzebyk
ABSTRACT
Understanding the conditions leading to harmful algal blooms, especially those produced by toxic dinoflagellate species, is important for environmental and health safety. In addition to investigations into the environmental conditions necessary for the formation of toxic blooms, we postulate that investigating gene expression in proliferating cells is essential for understanding bloom dynamics. Expressed sequence tags were produced from cultured cells of the toxic dinoflagellate Alexandrium catenella sampled during the initiation phase of growth using Sanger's method and by 454 pyrosequencing. A significant proportion of identified genes (ca. 25%) represented enzymes and proteins that participate in a variety of cellular regulatory mechanisms that may characterize proliferating cells, e.g., control of the cell cycle and division, regulation of transcription, translation and posttranslational protein modifications, signaling, intracellular trafficking, and transport. All of the several genes selected for gene expression assays due to their involvement in metabolism and the cell cycle were overexpressed during exponential growth. These data will be useful for investigating the mechanisms underlying growth and toxin production in toxic Alexandrium species and for studying and monitoring the development of toxic blooms.
Sigma-Aldrich
Terrific Broth, EZMix powder microbial growth medium
Pricing and availability is not currently available.