Merck
  • CD155/PVR enhances glioma cell dispersal by regulating adhesion signaling and focal adhesion dynamics.
CD155/PVR enhances glioma cell dispersal by regulating adhesion signaling and focal adhesion dynamics.
Cancer research (2005-12-03)
Kevin E Sloan, Jean K Stewart, Allison F Treloar, Russell T Matthews, Daniel G Jay
ABSTRACT
We recently identified the immunoglobulin-CAM CD155/PVR (the poliovirus receptor) as a regulator of cancer invasiveness and glioma migration, but the mechanism through which CD155/PVR controls these processes is unknown. Here, we show that expression of CD155/PVR in rat glioma cells that normally lack this protein enhances their dispersal both in vitro and on primary brain tissue. CD155/PVR expression also reduced substrate adhesion, cell spreading, focal adhesion density, and the number of actin stress fibers in a substrate-dependent manner. Furthermore, we found that expression of CD155/PVR increased Src/focal adhesion kinase signaling in a substrate-dependent manner, enhancing the adhesion-induced activation of paxillin and p130Cas in cells adhering to vitronectin. Conversely, depletion of endogenous CD155/PVR from human glioma cells inhibited their migration, increased cell spreading, and down-regulated the same signaling pathway. These findings implicate CD155/PVR as a regulator of adhesion signaling and suggest a pathway through which glioma and other cancer cells may acquire a dispersive phenotype.
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid
Sigma-Aldrich
Anti-Vinculin Antibody, clone VIIF9 (7F9), clone VIIF9 (7F9), Chemicon®, from mouse