Merck
  • Effect of N-2-mercaptopropionyl glycine on exercise-induced cardiac adaptations.
Effect of N-2-mercaptopropionyl glycine on exercise-induced cardiac adaptations.
American journal of physiology. Regulatory, integrative and comparative physiology (2011-02-11)
Matthew J Nelson, M Brennan Harris, Marvin O Boluyt, Hyun Seok Hwang, Joseph W Starnes
ABSTRACT
The purpose of this study was to test the hypothesis that exercise-induced cardiac adaptations would be attenuated by the free radical scavenger N-2-mercaptopropionyl glycine (MPG). Male Sprague-Dawley rats were divided into four groups (n = 9-13 per group) for 3-4 wk: sedentary (S), S+MPG (100 mg/kg ip daily), exercised on a treadmill (E) (60 min/day, 5 days/wk, at a speed of 20 m/min up a 6° grade in a 6°C room), or E+MPG given 10 min prior to exercise. Additional rats (n = 55) were used to determine acute exercise effects on myocardial redox state [nonprotein nonglutathione sulfhydryls (NPNGSH)] and PI3K/Akt signaling pathway activation. Compared with S, NPNGSH levels were 48% lower in E (P < 0.05) and unchanged in E+MPG (P > 0.05). MPG also attenuated exercise-induced activation of the signaling proteins Akt and S6. Hearts from the 4-wk groups were weighed, and cardiac function was evaluated using an isolated perfused working heart preparation. Similar increases (P < 0.05) in both exercised groups were observed for heart weight and heart weight-to-body weight ratio. Cardiac function improved in E vs. S, as indicated by greater (P < 0.05) external work performed (cardiac output × systolic pressure) and efficiency of external work (work/Vo(2)). MPG prevented these exercise-induced functional improvements. Skeletal muscle mitochondria content increased to similar levels in E and E+MPG. This study provides evidence that free radicals do not play an essential role in the development of exercise-induced cardiac hypertrophy; however, they appear to be involved in functional cardiac adaptations, which may be mediated through the PI3K/Akt pathway.
Sigma-Aldrich
Anti-phospho-Akt1/PKBα (Ser473) Antibody, clone SK703, rabbit monoclonal, clone SK703, Upstate®, from rabbit
Pricing and availability is not currently available.