Merck
  • Dual roles of sphingolipids in signaling of the escape from and onset of apoptosis in a mouse cytotoxic T-cell line, CTLL-2.
Dual roles of sphingolipids in signaling of the escape from and onset of apoptosis in a mouse cytotoxic T-cell line, CTLL-2.
The Journal of biological chemistry (1996-01-19)
S Nakamura, Y Kozutsumi, Y Sun, Y Miyake, T Fujita, T Kawasaki
ABSTRACT
In our previous study, the sphingosine-like immunosuppressant, ISP-1, was found to suppress the proliferation of an interleukin-2-dependent cytotoxic T cell line, CTLL-2, through the inhibition of serine palmitoyltransferase, which catalyzes the committed step of sphingolipid biosynthesis. Analysis of the effect of ISP-1 by flow cytometry revealed that the ISP-1-dependent decrease in cell number was not due to inhibition of the cell cycle progression of CTLL-2 cells but to the induction of apoptosis of the cells. The ISP-1-induced apoptosis was inhibited by the addition of sphingosine (2 microM), suggesting that this ISP-1-induced apoptosis is triggered by the decrease in the intracellular levels of sphingolipids caused by the inhibition of serine palmitoyltransferase. However, another interleukin-2-dependent cell line, F7, which was derived from a mouse pro-B cell line, did not show ISP-1-dependent apoptosis, indicating that the effect of ISP-1 may be specific for a certain type of T cell lineage such as CTLL-2. On the other hand, a high dose of sphingosine (5 microM) by itself induced the apoptosis of CTLL-2 cells. This sphingosine-dependent apoptosis was also observed with F7 cells. These results provide evidence that the intracellular levels of sphingolipids play an important role in the signaling of the escape from and onset of apoptosis of CTLL-2 cells.
Sigma-Aldrich
Myriocin, Mycelia sterilia, A potent immunosuppressant.
Pricing and availability is not currently available.
Sigma-Aldrich
Myriocin from Mycelia sterilia, ≥98% (HPLC), powder
Pricing and availability is not currently available.