Merck
Homeobox gene Dlx-2 is implicated in metabolic stress-induced necrosis.
Molecular cancer (2011-09-16)
Su Yeon Lee, Hyun Min Jeon, Cho Hee Kim, Min Kyung Ju, Hye Sun Bae, Hye Gyeong Park, Sung-Chul Lim, Song Iy Han, Ho Sung Kang
ABSTRACT
In contrast to tumor-suppressive apoptosis and autophagic cell death, necrosis promotes tumor progression by releasing the pro-inflammatory and tumor-promoting cytokine high mobility group box 1 (HMGB1), and its presence in tumor patients is associated with poor prognosis. Thus, necrosis has important clinical implications in tumor development; however, its molecular mechanism remains poorly understood. In the present study, we show that Distal-less 2 (Dlx-2), a homeobox gene of the Dlx family that is involved in embryonic development, is induced in cancer cell lines dependently of reactive oxygen species (ROS) in response to glucose deprivation (GD), one of the metabolic stresses occurring in solid tumors. Increased Dlx-2 expression was also detected in the inner regions, which experience metabolic stress, of human tumors and of a multicellular tumor spheroid, an in vitro model of solid tumors. Dlx-2 short hairpin RNA (shRNA) inhibited metabolic stress-induced increase in propidium iodide-positive cell population and HMGB1 and lactate dehydrogenase (LDH) release, indicating the important role(s) of Dlx-2 in metabolic stress-induced necrosis. Dlx-2 shRNA appeared to exert its anti-necrotic effects by preventing metabolic stress-induced increases in mitochondrial ROS, which are responsible for triggering necrosis. These results suggest that Dlx-2 may be involved in tumor progression via the regulation of metabolic stress-induced necrosis.
Sigma-Aldrich
Anti-Dlx2 Antibody, Chemicon®, from rabbit
Pricing and availability is not currently available.