Merck
  • The Role of GLP1 in Rat Steatotic and Non-Steatotic Liver Transplantation from Cardiocirculatory Death Donors.
The Role of GLP1 in Rat Steatotic and Non-Steatotic Liver Transplantation from Cardiocirculatory Death Donors.
Cells (2019-12-15)
Cindy G Avalos-de León, Mónica B Jiménez-Castro, María Eugenia Cornide-Petronio, Araní Casillas-Ramírez, Carmen Peralta
ABSTRACT
In liver transplantation (LT), organ shortage has led to the use of steatotic and non-steatotic grafts from donors after cardiocirculatory death (DCD). However, these grafts, especially those with steatosis, exhibit poor post-operative outcomes. To address this problem, we investigated the roles of gut-derived glucagon-like peptide 1 (GLP1) and dipeptidyl peptidase 4 (DPP4), the serine protease that cleaves it, in steatotic and non-steatotic LT from DCDs. Using Zucker rats, liver grafts from DCDs were cold stored and transplanted to recipients. GLP1 was administered to donors. The levels of GLP1 in intestine and of both GLP1 and DDP4 in circulation were unaltered following cardiocirculatory death (CD). In steatotic livers from DCD, increased GLP1 and decreased DPP4 were recorded, and administration of GLP1 caused a rise in hepatic GLP1 and a reduction in DDP4. This protected against inflammation, damage, and proliferation failure. Conversely, low GLP1 and high DDP4 were observed in non-steatotic livers from DCD. The exogenous GLP1 did not modify hepatic DDP4, and the accumulated GLP1 exerted harmful effects, increasing damage, inflammation, and regeneration failure. Herein, we show that there are differences in GLP1/DDP4 regulation depending on the type of liver implanted, suggesting that GLP1 can be used as a novel and effective therapy in steatotic grafts from DCDs but that it is not appropriate for non-steatotic DCDs.
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, purified immunoglobulin, buffered aqueous solution