Merck
  • Temporary conductive hearing loss in early life impairs spatial memory of rats in adulthood.
Temporary conductive hearing loss in early life impairs spatial memory of rats in adulthood.
Brain and behavior (2018-06-02)
Han Zhao, Li Wang, Liang Chen, Jinsheng Zhang, Wei Sun, Richard J Salvi, Yi-Na Huang, Ming Wang, Lin Chen
ABSTRACT
It is known that an interruption of acoustic input in early life will result in abnormal development of the auditory system. Here, we further show that this negative impact actually spans beyond the auditory system to the hippocampus, a system critical for spatial memory. We induced a temporary conductive hearing loss (TCHL) in P14 rats by perforating the eardrum and allowing it to heal. The Morris water maze and Y-maze tests were deployed to evaluate spatial memory of the rats. Electrophysiological recordings and anatomical analysis were made to evaluate functional and structural changes in the hippocampus following TCHL. The rats with the TCHL had nearly normal hearing at P42, but had a decreased performance with the Morris water maze and Y-maze tests compared with the control group. A functional deficit in the hippocampus of the rats with the TCHL was found as revealed by the depressed long-term potentiation and the reduced NMDA receptor-mediated postsynaptic current. A structural deficit in the hippocampus of those animals was also found as revealed the abnormal expression of the NMDA receptors, the decreased number of dendritic spines, the reduced postsynaptic density and the reduced level of neurogenesis. Our study demonstrates that even temporary auditory sensory deprivation in early life of rats results in abnormal development of the hippocampus and consequently impairs spatial memory in adulthood.
Sigma-Aldrich
Anti-GAPDH Antibody, from chicken, purified by affinity chromatography
Sigma-Aldrich
Anti-NR2A Antibody, clone A12W, rabbit monoclonal
Sigma-Aldrich
Anti-NMDAR2B Antibody, ascites fluid, clone 1E4.25A4, Chemicon®