Merck
  • FANCC localizes with UNC5A at neurite outgrowth and promotes neuritogenesis.
FANCC localizes with UNC5A at neurite outgrowth and promotes neuritogenesis.
BMC research notes (2018-09-15)
FengFei Huang, Manel Ben Aissa, Georges Lévesque, Madeleine Carreau
ABSTRACT
The Uncoordinated 5A (UNC5A) protein is part of a family of receptors that play roles in axonal pathfinding and cell migration. We previously showed that the Fanconi anemia C protein (FANCC) interacts with UNC5A and delays UNC5A-mediated apoptosis. FANCC is a predominantly cytoplasmic protein that has multiple functions including DNA damage signaling, oxygen radical metabolism, signal transduction, transcriptional regulation and apoptosis. Given the direct interaction between FANCC and UNC5A and that FANCC interferes with UNC5A-mediated apoptosis, we explored the possibility that FANCC might play a role in axonal-like growth processes. Here we show that FANCC and UNC5A are localized to regions of neurite outgrowth during neuronal cell differentiation. We also show that absence of FANCC is required for neurite outgrowth. In addition, FANCC seems required for UNC5A expression. Results from this study combined with our previous report suggest that FANCC plays a role in tissue development through the regulation of UNC5A-mediated functions.
Sigma-Aldrich
Anti-FANCC Antibody, clone 8F3, clone 8F3, 1 mg/mL, from mouse
Sigma-Aldrich
Anti-α-Tubulin antibody, Mouse monoclonal, clone DM1A, purified from hybridoma cell culture