Merck
Organic Mechanoluminescence with Aggregation-Induced Emission.
Chemistry, an Asian journal (2018-08-10)
Eethamukkala Ubba, Yu Tao, Zhiyong Yang, Juan Zhao, Leyu Wang, Zhenguo Chi
ABSTRACT
Mechanoluminescence (ML) involves the emission of light from various organic and inorganic materials upon mechanical stimulus. In this review, we contemplate the advances of metal-free, purely organic ML compounds with aggregation-induced emission (AIE), which give strong emission upon mechanical force. We show the new ML-AIE based materials can be used explore the basic of ML properties with structure-property relationships and molecular interactions. The flourishing idea of ML-AIE may help to find the characteristic and elusive component of ML as it partners with AIE, which upgrades the emission property in solids or aggregates. Recent advances in ML-AIE information could increase the wide scope of research in this expanding area, as these materials have promising applications in different optoelectronic areas.
Sigma-Aldrich
HNMB, ≥98%
Pricing and availability is not currently available.