Merck
  • A monoclonal anti-interleukin 8 antibody (WS-4) inhibits cytokine response and acute lung injury in experimental severe acute necrotising pancreatitis in rabbits.
A monoclonal anti-interleukin 8 antibody (WS-4) inhibits cytokine response and acute lung injury in experimental severe acute necrotising pancreatitis in rabbits.
Gut (1999-04-06)
M O Osman, J U Kristensen, N O Jacobsen, S B Lausten, B Deleuran, M Deleuran, B Gesser, K Matsushima, C G Larsen, S L Jensen
ABSTRACT
Interleukin 8 (IL-8) has recently been proposed to have an important role in mediating the development of the systemic sequelae associated with severe acute pancreatitis. To define the role of IL-8 in acute pancreatitis by neutralising its effects with a monoclonal anti-IL-8 antibody (WS-4), in a rabbit model of severe acute pancreatitis. Acute pancreatitis was induced by retrograde injection of 5% chenodeoxycholic acid into the pancreatic duct and duct ligation. Twenty rabbits were divided equally into two groups: acute pancreatitis controls received physiological saline and the treated group received WS-4, 30 minutes before induction of acute pancreatitis. Pretreatment of animals with WS-4 resulted in significant down regulation of serum IL-8 and tumour necrosis factor alpha (TNF-alpha) from three to six hours after induction of acute pancreatitis (p = 0.011 and 0.047 for IL-8 and 0.033 and 0.022 for TNF-alpha, respectively). In addition, a significant reduction in the CD11b and CD18 positive cells and the amount of interstitial neutrophil infiltration in the lungs from WS-4 treated animals was seen. In contrast, WS-4 did not alter the amount of pancreatic necrosis and the serum concentrations of amylase, lipase, calcium, and glucose. WS-4 cannot change the amount of pancreatic necrosis induced by injection of 5% bile acid, but does reduce the acute lung injury, presumably through inhibition of circulating IL-8 and TNF-alpha, and CD11b/CD18 in lung tissue. Therefore, a role of IL-8 in the progression of acute pancreatitis and the development of its systemic complications is suggested.
Sigma-Aldrich
Chenodeoxycholic acid
Pricing and availability is not currently available.