Merck
  • Relevant assay to study the adhesion of Plasmodium falciparum-infected erythrocytes to the placental epithelium.
Relevant assay to study the adhesion of Plasmodium falciparum-infected erythrocytes to the placental epithelium.
PloS one (2011-07-07)
Philippe Boeuf, Wina Hasang, Eric Hanssen, Jocelyn D Glazier, Stephen J Rogerson
ABSTRACT
In placental malaria, Plasmodium falciparum-infected erythrocytes adhere to the apical plasma membrane of the placental epithelium, triggering an impairment of placental function detrimental to the fetus. The design of anti-adhesion intervention strategies requires a detailed understanding of the mechanisms involved. However, most adhesion assays lack in vivo relevance and are hardly quantitative. Here, we describe a flow cytometry-based adhesion assay that is fully relevant by using apical epithelial plasma membrane vesicles as the adhesion matrix, and being applicable to infected erythrocytes directly isolated from patients. Adhesion is measured both as the percentage of pathogens bound to epithelial membrane vesicles as well as the mean number of vesicles bound per infected erythrocytes. We show that adhesins alternative to those currently identified could be involved. This demonstrates the power of this assay to advance our understanding of epithelial adhesion of infected erythrocytes and in the design of intervention strategies.
Sigma-Aldrich
Chondroitin sulfate A sodium salt from bovine trachea, lyophilized powder, BioReagent, suitable for cell culture
Pricing and availability is not currently available.
Sigma-Aldrich
Chondroitinase ABC from Proteus vulgaris, BSA free, lyophilized powder, specific activity 50-250 units/mg protein
Pricing and availability is not currently available.